Tricosanoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

T6543-100MG T6543-1G
SIALT6543-100MGEA 113 GBP
SIALT6543-100MG SIALT6543-1G
Tricosanoic acid, Sigma-Aldrich®
Tricosanoic acid
Formula: C₂₃H₄₆O₂
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002808
CAS Number: 2433-96-7
EINECS: 219-419-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR