Stearic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
175366-100G
175366-1KG
175366-5KG
SIAL175366-100GEA
11.5
GBP
SIAL175366-100G
SIAL175366-1KG
SIAL175366-5KG
Stearic acid, Sigma-Aldrich®
Stearic acid
Formula:
C₁₈H₃₆O₂ Boiling Pt: 361 °C (1013 hPa) Melting Pt: 67 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 196 °C (closed cup) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002752 CAS Number: 57-11-4 EINECS: 200-313-4 Merck Index: 13,08882 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...