Nonadecanoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Nonadecylic acid

M-Clarity™ quality level = MQ200

N5252-1G N5252-5G
SIALN5252-1GEA 129 GBP
SIALN5252-1G SIALN5252-5G
Nonadecanoic acid, Sigma-Aldrich®
Nonadecanoic acid
Formula: C₁₉H₃₈O₂
MW: 298.51 g/mol
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 0.8468 g/cm³ (70 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002754
CAS Number: 646-30-0
EINECS: 211-472-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR