Capric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ300
W236403-10KG-K
W236403-1KG-K
W236403-25KG-K
W236403-5KG-K
SIALW236403-10KG-KEA
536
GBP
SIALW236403-10KG-K
SIALW236403-1KG-K
SIALW236403-25KG-K
SIALW236403-5KG-K
Capric acid, Sigma-Aldrich®
Capric acid
Formula:
C₁₀H₂₀O₂ Boiling Pt: 268 °C (1013 hPa) Melting Pt: 29…32 °C Density: 0.893 g/cm³ (20 °C) Flash Pt: 147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004441 CAS Number: 334-48-5 EINECS: 206-376-4 Merck Index: 13,01764 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...