Sebacoyl dichloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Sebacoyl chloride

M-Clarity™ quality level = MQ200

84850-100ML 84850-25ML 84850-500ML
SIAL84850-100MLEA 160 GBP
SIAL84850-100ML SIAL84850-25ML SIAL84850-500ML
Sebacoyl dichloride, Sigma-Aldrich®
Sebacoyl dichloride
Formula: C₁₀H₁₆Cl₂O₂
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR