1-Naphthyl acetate
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Naphthalen-1-yl acetate, Ethanoic acid 1-naphthyl ester, Acetic acid 1-naphthyl ester, α-Naphthyl acetate, Acetic acid α-naphthyl ester, Ethanoic acid α-naphthyl ester
30040.01
30040.02
SERA30040.01EA
76.88
GBP
SERA30040.01
SERA30040.02
1-Naphthyl acetate
1-Naphthyl acetate
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol Boiling Pt: 310 °C (1013 hPa) Melting Pt: 41…43 °C Density: 1,1305 g/cm³ (50 °C) Flash Pt: >100 °C Storage Temperature: Freezer |
MDL Number:
MFCD00003922 CAS Number: 830-81-9 EINECS: 212-599-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...