Dimethyl malonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester
M-Clarity™ quality level = MQ100
63380-1L
63380-250ML
SIAL63380-1LEA
149
GBP
SIAL63380-1L
SIAL63380-250ML
Dimethyl malonate, Sigma-Aldrich®
Dimethyl malonate
Formula:
C₅H₈O₄ Boiling Pt: 181 °C (1013 hPa) Melting Pt: –62 °C Density: 1.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008460 CAS Number: 108-59-8 EINECS: 203-597-8 Merck Index: 12,06169 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...