Nonyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

W278807-1KG W278807-4KG W278807-SAMPLE
SIALW278807-1KGEA 196 GBP
SIALW278807-1KG SIALW278807-4KG SIALW278807-SAMPLE
Nonyl acetate, Sigma-Aldrich®
Nonyl acetate
Formula: C₁₁H₂₂O₂
Boiling Pt: 208…212 °C (1013 hPa)
Melting Pt: –26 °C
Density: 0.868 g/cm³ (20 °C)
MDL Number: MFCD00027340
CAS Number: 143-13-5
EINECS: 205-585-8
Merck Index: 12,06773

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR