Heptyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

W254703-1KG-K W254703-4KG-K W254703-9KG-K
SIALW254703-1KG-KEA 81.8 GBP
SIALW254703-1KG-K SIALW254703-4KG-K SIALW254703-9KG-K
Heptyl acetate, Sigma-Aldrich®
Heptyl acetate
Formula: C₉H₁₈O₂
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –50.2 °C
Density: 0.8891 g/cm³ (0 °C)
MDL Number: MFCD00027311
CAS Number: 112-06-1
EINECS: 203-932-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR