Isobutyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ300

W217506-1KG-K W217506-20KG-K W217506-9KG-K
SIALW217506-1KG-KEA 79.2 GBP
SIALW217506-1KG-K SIALW217506-20KG-K SIALW217506-9KG-K
Isobutyl acetate, Sigma-Aldrich®
Isobutyl acetate
Formula: C₆H₁₂O₂
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008932
CAS Number: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3,II
Merck Index: 13,05147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR