Methyl laurate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

M-Clarity™ quality level = MQ300

W271500-1KG W271500-4KG W271500-9KG W271500-SAMPLE
SIALW271500-1KGEA 104 GBP
SIALW271500-1KG SIALW271500-4KG SIALW271500-9KG SIALW271500-SAMPLE
Methyl laurate, Sigma-Aldrich®
Methyl laurate
Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266
Melting Pt: 4.5
Density: 0.8694 g/cm³ (20 °C)
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR