Ethyl laurate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ300
W244104-1KG-K
W244104-20KG-K
W244104-4KG-K
SIALW244104-1KG-KEA
80.5
GBP
SIALW244104-1KG-K
SIALW244104-20KG-K
SIALW244104-4KG-K
Ethyl laurate, Sigma-Aldrich®
Ethyl laurate
Formula:
C₁₄H₂₈O₂ Boiling Pt: 269 °C (1013 hPa) Melting Pt: –2 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015065 CAS Number: 106-33-2 EINECS: 203-386-0 Merck Index: 13,03851 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...