4'-Aminoacetanilide 95%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: N-(4-Aminophenyl)acetamide

400600250 400601000
ACRO400600250EA 38 GBP
ACRO400600250 ACRO400601000
4'-Aminoacetanilide 95%
4'-Aminoacetanilide
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 267 °C
Melting Pt: 159…165 °C
Density: 1,27 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007853
CAS Number: 122-80-5
EINECS: 204-576-6
Merck Index: 13,00410

Order Now

Specification Test Results

Appearance Pink to brown Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 165 °C
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR