Methylurea ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B24772.0B
B24772.30
B24772.0BEA
105
GBP
B24772.0B
B24772.30
Methylurea ≥98%
Methylurea
Purity: 98%
CAS No.: 598-50-5
Molecular Formula: CH3NHCONH2
Formula Weight: 74.08
Odour: Odourless
pH: 6.7
Melting Point: 96-101°
Beilstein Reference: 878189
MDL No.: MFCD00007950
Formula:
C₂H₆N₂O Boiling Pt: 240 °C (1013 hPa) Melting Pt: 96…101 °C Density: 1.204 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007950 CAS Number: 598-50-5 EINECS: 209-935-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...