Methylurea 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

127991000 127995000
ACRO127991000EA 17.3 GBP
ACRO127991000 ACRO127995000
Methylurea 97%
Methylurea
Formula: C₂H₆N₂O
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 98…102 °C
Density: 1,204 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007950
CAS Number: 598-50-5
EINECS: 209-935-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Water ≤2 %
HPLC ≥96.0 % (on dry substance)
Titration with HClO4 ≥96.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR