ADA, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
M-Clarity™ quality level = MQ200
00307-100G
00307-25G
00307-5G
SIAL00307-100GEA
405
GBP
SIAL00307-100G
SIAL00307-25G
SIAL00307-5G
ADA, Sigma-Aldrich®
ADA
Formula:
H₂NC(O)CH₂N(CH₂COOH)₂ MW: 190.16 g/mol Melting Pt: 217…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008031 CAS Number: 26239-55-4 EINECS: 247-530-0 Merck Index: 13,00148 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...