Acetyl bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

135968-100G 135968-500G
SIAL135968-100GEA 106 GBP
SIAL135968-100G SIAL135968-500G
Acetyl bromide, Sigma-Aldrich®
Acetyl bromide
Formula: C₂H₃BrO
MW: 122.95 g/mol
Boiling Pt: 77 °C (1013 hPa)
Melting Pt: –96 °C
Density: 1.65 g/cm³ (20 °C)
Flash Pt: >71 °C
MDL Number: MFCD00000114
CAS Number: 506-96-7
EINECS: 208-061-7
UN: 1716
ADR: 8,II
Merck Index: 13,00084

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR