Hydroxyacetone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

138185-100G 138185-500G 138185-5G
SIAL138185-100GEA 59.8 GBP
SIAL138185-100G SIAL138185-500G SIAL138185-5G
Hydroxyacetone, Sigma-Aldrich®
Hydroxyacetone
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.0928 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004669
CAS Number: 116-09-6
EINECS: 204-124-8
UN: 1224
ADR: 3,III
Merck Index: 13,00066

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR