Biuret 97%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Carbamylurea, Allophanic acid amide

106520050 106520250 106521000
ACRO106520050EA 18.9 GBP
ACRO106520050 ACRO106520250 ACRO106521000
Biuret 97%, extra pure
Biuret
Formula: C₂H₅N₃O₂
Melting Pt: 187…194 °C (dec.)
Density: 1,467 g/cm³ (10 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007946
CAS Number: 108-19-0
EINECS: 203-559-0
Merck Index: 13,01305
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da007a12828859edd80cac6339b70b729a53acc4&vid=58b4eefca29a4ffb12285d8909475d80ef5ff811&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 184 to 190 °C
Assay ≥96.0 % (ex N)
Loss on drying ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR