2-Undecanone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl nonyl ketone, Nonyl methyl ketone

M-Clarity™ quality level = MQ300

W309303-1KG-K W309303-250G-K W309303-4KG-K W309303-8KG-K
SIALW309303-1KG-KEA 212 GBP
SIALW309303-1KG-K SIALW309303-250G-K SIALW309303-4KG-K SIALW309303-8KG-K
2-Undecanone, Sigma-Aldrich®
2-Undecanone
Formula: C₁₁H₂₂O
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: ∼12 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00009583
CAS Number: 112-12-9
EINECS: 203-937-5
UN: 3082
ADR: 9,III
Merck Index: 12,06178

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR