2-Tridecanone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


MQ300

W338818-100G-K W338818-1KG-K W338818-4KG-K
SIALW338818-100G-KEA 78 GBP
SIALW338818-100G-K SIALW338818-1KG-K SIALW338818-4KG-K
2-Tridecanone, Sigma-Aldrich®
2-Tridecanone
Formula: C₁₃H₂₆O
MW: 198.35 g/mol
Boiling Pt: 263
Melting Pt: 25…27 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008968
CAS Number: 593-08-8
EINECS: 209-784-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR