Chloroacetic anhydride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

215163-100G 215163-25G
SIAL215163-100GEA 190 GBP
SIAL215163-100G SIAL215163-25G
Chloroacetic anhydride, Sigma-Aldrich®
Chloroacetic anhydride
Formula: C₄H₄Cl₂O₃
MW: 170.98 g/mol
Boiling Pt: 203
Melting Pt: 56…58 °C
Density: 1.5497 g/cm³ (20 °C)
MDL Number: MFCD00000929
CAS Number: 541-88-8
EINECS: 208-794-2
UN: 3261
ADR: 8 II
Merck Index: 13,02130

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR