Chloroiodomethane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

242861-25G 242861-5G
SIAL242861-25GEA 88.2 GBP
SIAL242861-25G SIAL242861-5G
Chloroiodomethane, Sigma-Aldrich®
Chloroiodomethane
Formula: CH₂ClI
Boiling Pt: 108 °C (1013 hPa)
Density: 2.416 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001078
CAS Number: 593-71-5
EINECS: 209-804-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR