1-Chloro-2-iodoethane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001485-5G 001485-25G
FLUO001485-5GEA 0 GBP
FLUO001485-5G FLUO001485-25G
1-Chloro-2-iodoethane
1-Chloro-2-iodoethane
Formula: C₂H₄ClI
Boiling Pt: 61…62 °C (30 mmHg)
Melting Pt: –15.6 °C
Density: 2,088 g/cm³ (20 °C)
MDL Number: MFCD00041453
CAS Number: 624-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR