Chlorodifluoroacetic anhydride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002452-25G 002452-5G
FLUO002452-25GEA 0 GBP
FLUO002452-25G FLUO002452-5G
Chlorodifluoroacetic anhydride
Chlorodifluoroacetic anhydride
Formula: C₄Cl₂F₄O₃
Boiling Pt: 93.5…94 °C (1013 hPa)
Density: 1.3953 g/cm³ (20 °C)
MDL Number: MFCD00000774
CAS Number: 2834-23-3
EINECS: 220-603-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR