Chlorodifluoroacetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Difluorochloroacetic acid

003151-25G 003151-100G 003151-500G
FLUO003151-25GEA 0 GBP
FLUO003151-25G FLUO003151-100G FLUO003151-500G
Chlorodifluoroacetic acid
Chlorodifluoroacetic acid
Formula: C₂HClF₂O₂
MW: 130.48 g/mol
Boiling Pt: 122 °C (1013 hPa)
Density: 2.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004176
CAS Number: 76-04-0
EINECS: 200-928-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR