Ethyl chlorodifluoroacetate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic ethyl ester
001174-25G
001174-100G
FLUO001174-25GEA
0
GBP
FLUO001174-25G
FLUO001174-100G
Ethyl chlorodifluoroacetate
Ethyl chlorodifluoroacetate
Formula:
C₄H₅ClF₂O₂ Boiling Pt: 95…96 °C (1013 hPa) Density: 1.253 g/cm³ (20 °C) |
MDL Number:
MFCD00013662 CAS Number: 383-62-0 EINECS: 206-850-0 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...