2-Chloro-4'-fluoroacetanilide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004368-5G 004368-25G 004368-100G
FLUO004368-5GEA 0 GBP
FLUO004368-5G FLUO004368-25G FLUO004368-100G
2-Chloro-4'-fluoroacetanilide
2-Chloro-4'-fluoroacetanilide
Formula: C₈H₇ClFNO
MDL Number: MFCD00015730
CAS Number: 351-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR