Valeraldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Pentylaldehyde, Amylaldehyde, Pentanal
M-Clarity™ quality level = MQ300
W309818-1KG-K
W309818-20KG-K
W309818-4KG-K
W309818-8KG-K
SIALW309818-1KG-KEA
89.8
GBP
SIALW309818-1KG-K
SIALW309818-20KG-K
SIALW309818-4KG-K
SIALW309818-8KG-K
Valeraldehyde, Sigma-Aldrich®
Valeraldehyde
Formula:
C₅H₁₀O Boiling Pt: 103 °C (1013 hPa) Melting Pt: –92 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: 7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007026 CAS Number: 110-62-3 EINECS: 203-784-4 UN: 2058 ADR: 3,II Merck Index: 12,10040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...