Valeraldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Pentylaldehyde, Amylaldehyde, Pentanal

M-Clarity™ quality level = MQ100

110132-1L 110132-250ML
SIAL110132-1LEA 150 GBP
SIAL110132-1L SIAL110132-250ML
Valeraldehyde, Sigma-Aldrich®
Valeraldehyde
Formula: C₅H₁₀O
Boiling Pt: 103 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007026
CAS Number: 110-62-3
EINECS: 203-784-4
UN: 2058
ADR: 3,II
Merck Index: 12,10040

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR