2-Bromoethanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

B65586-100G B65586-500G B65586-5G
SIALB65586-100GEA 104 GBP
SIALB65586-100G SIALB65586-500G SIALB65586-5G
2-Bromoethanol, Sigma-Aldrich®
2-Bromoethanol
Formula: BrH₂CCH₂OH
Boiling Pt: 149 °C (1013 hPa)
Melting Pt: –80 °C
Density: 1.76 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002827
CAS Number: 540-51-2
EINECS: 208-748-1
UN: 2927
ADR: 6.1,II
Merck Index: 13,03826

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR