1-Bromoperfluorohexane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002309-25G 002309-100G
FLUO002309-25GEA 0 GBP
FLUO002309-25G FLUO002309-100G
1-Bromoperfluorohexane
1-Bromoperfluorohexane
Formula: BrF₂C(CF₂)₄CF₃
MDL Number: MFCD00042349
CAS Number: 335-56-8
EINECS: 206-391-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR