1-Bromoperfluorooctane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002311-100G 002311-500G 002311-25G 002311-5G
FLUO002311-100GEA 0 GBP
FLUO002311-100G FLUO002311-500G FLUO002311-25G FLUO002311-5G
1-Bromoperfluorooctane
1-Bromoperfluorooctane
Formula: BrF₂C(CF₂)₆CF₃
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.918 g/cm³ (25 °C)
MDL Number: MFCD00042082
CAS Number: 423-55-2
EINECS: 207-028-4
Merck Index: 12,07299

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR