6-Bromocaproic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
6-Bromohexanoic acid
002967-25G
002967-100G
002967-10G
FLUO002967-25GEA
0
GBP
FLUO002967-25G
FLUO002967-100G
FLUO002967-10G
6-Bromocaproic acid
6-Bromocaproic acid
Formula:
BrH₂C(CH₂)₄COOH Boiling Pt: 165 °C (1013 hPa) Melting Pt: 31…34 °C Density: 1.4068 g/cm³ (30 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004422 CAS Number: 4224-70-8 EINECS: 224-176-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...