Ethyl-5-bromovalerate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromovaleric acid ethyl ester, Ethyl 5-bromovalerate

M-Clarity™ quality level = MQ200

129100-100G 129100-25G 129100-5G
SIAL129100-100GEA 200 GBP
SIAL129100-100G SIAL129100-25G SIAL129100-5G
Ethyl-5-bromovalerate, Sigma-Aldrich®
Ethyl 5-bromovalerate
Formula: BrH₂C(CH₂)₃COOCH₂CH₃
MW: 209.08 g/mol
Boiling Pt: 104 °C (1013 hPa)
Density: 1.2849 g/cm³ (20 °C)
MDL Number: MFCD00000266
CAS Number: 14660-52-7
EINECS: 238-705-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR