Bromoacetamide ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Bromoacetamide, beta-Bromoacetamide

A11926.14 A11926.06
A11926.14EA 99.3 GBP
A11926.14 A11926.06
Bromoacetamide ≥98%
Bromoacetamide

Purity: 98% CAS No.: 683-57-8 Molecular Formula: C2H4BrNO Formula Weight: 137.97 Melting Point: 87-90° Beilstein Reference: 1739073 UN No.: 2923 MDL No.: MFCD00008025

Formula: BrCH₂CONH₂
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008025
CAS Number: 683-57-8
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR