Bromoacetamide 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-Bromoacetamide, beta-Bromoacetamide
291100250
291100050
ACRO291100250EA
119
GBP
ACRO291100250
ACRO291100050
Bromoacetamide 98%
Bromoacetamide
Formula:
BrCH₂CONH₂ Boiling Pt: 241 °C (1013 hPa) Melting Pt: 88…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008025 CAS Number: 683-57-8 UN: 2923 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 87 to 91 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...