6-Bromohexanoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003076-25G 003076-100G
FLUO003076-25GEA 0 GBP
FLUO003076-25G FLUO003076-100G
6-Bromohexanoyl chloride
6-Bromohexanoyl chloride
Formula: BrH₂C(CH₂)₄COOCl
MW: 213.5 g/mol
Boiling Pt: 130
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000761
CAS Number: 22809-37-6
EINECS: 245-236-7
UN: 3265

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR