Bromoacetyl bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

B56412-100G B56412-500G B56412-5G
SIALB56412-100GEA 74.5 GBP
SIALB56412-100G SIALB56412-500G SIALB56412-5G
Bromoacetyl bromide, Sigma-Aldrich®
Bromoacetyl bromide
Formula: BrCH₂COOBr
Boiling Pt: 148 °C (1013 hPa)
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000115
CAS Number: 598-21-0
EINECS: 209-923-5
UN: 2513
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR