Methyl bromopyruvate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-Bromopyruvate, Methyl 3-Bromo-2-oxopropionate, Bromopyruvic acid methyl ester, 3-Bromopyruvic acid methyll ester, 3-Bromo-2-oxopropionic acid methyll ester, 3-Bromo-2-oxopropionic acid methyl ester

16491-25ML 16491-5ML
SIAL16491-25MLEA 366 GBP
SIAL16491-25ML SIAL16491-5ML
Methyl bromopyruvate, Sigma-Aldrich®
Methyl bromopyruvate
Formula: BrH₂CC(O)COOCH₃
MW: 180.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862963
CAS Number: 7425-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR