Dibromoacetonitrile ≥94%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

A16994.06 A16994.14 A16994.22
A16994.06EA 19.8 GBP
A16994.06 A16994.14 A16994.22
Dibromoacetonitrile ≥94%
Dibromoacetonitrile

Purity: 94% CAS No.: 3252-43-5 Molecular Formula: C2HBr2N Formula Weight: 198.84 Flash Point: 56°(133°F) Refractive Index: 1.5420 Odour: Pungent, makes eyes water Boiling Point: 67-69°/24mm Beilstein Reference: 1739037 UN No.: 3286 MDL No.: MFCD00001856

Formula: Br₂CHCN
Boiling Pt: 67…69 °C (24 mmHg)
Density: 2.296
Flash Pt: 56 °C (133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001856
CAS Number: 3252-43-5
EINECS: 221-843-2
UN: 3286
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR