1-Bromotridecane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tridecyl bromide

MQ200

192422-25G 192422-5G
SIAL192422-25GEA 618 GBP
SIAL192422-25G SIAL192422-5G
1-Bromotridecane, Sigma-Aldrich®
1-Bromotridecane
Formula: BrH₂C(CH₂)₁₁CH₃
MW: 263.26 g/mol
Boiling Pt: 292
Melting Pt: 6
Density: 1.0262 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000227
CAS Number: 765-09-3
EINECS: 212-138-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR