L(-)-Fenchone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(1R)-(-)-Fenchone
M-Clarity™ quality level = MQ300
W507709-100G-K
W507709-1KG-K
W507709-5KG-K
SIALW507709-100G-KEA
75.9
GBP
SIALW507709-100G-K
SIALW507709-1KG-K
SIALW507709-5KG-K
L(-)-Fenchone, Sigma-Aldrich®
L(-)-Fenchone
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Boiling Pt: 192 °C (1013 hPa) Melting Pt: 8.5 °C Density: 0.962 g/cm³ (0 °C) |
MDL Number:
MFCD00151104 CAS Number: 7787-20-4 EINECS: 232-107-5 UN: 2717 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...