(R)-(+)-3-Butyne-2-ol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne

M-Clarity™ quality level = MQ100

464821-1G 464821-5G
SIAL464821-1GEA 175 GBP
SIAL464821-1G SIAL464821-5G
(R)-(+)-3-Butyne-2-ol, Sigma-Aldrich®
(R)-(+)-3-Butyne-2-ol
Formula: C₄H₆O
Boiling Pt: 103 °C (1013 hPa)
Density: 0.891 g/cm³ (20 °C)
MDL Number: MFCD00211237
CAS Number: 42969-65-3
UN: 2929
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR