L(-)-Tyrosine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
M-Clarity™ quality level = MQ300
W373605-100G
W373605-1KG
W373605-SAMPLE
SIALW373605-100GEA
75.8
GBP
SIALW373605-100G
SIALW373605-1KG
SIALW373605-SAMPLE
L(-)-Tyrosine, Sigma-Aldrich®
L(-)-Tyrosine
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Boiling Pt: 351 °C (1013 hPa) Melting Pt: 285 °C Density: 1.41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002606 CAS Number: 60-18-4 EINECS: 200-460-4 Merck Index: 13,09907 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...