(S)-(-)-2-Methyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
65980-100ML
65980-500ML
SIAL65980-100MLEA
106
GBP
SIAL65980-100ML
SIAL65980-500ML
(S)-(-)-2-Methyl-1-butanol, Sigma-Aldrich®
(S)-(-)-2-Methyl-1-butanol
Formula:
C₅H₁₂O Boiling Pt: 128 °C (1013 hPa) Melting Pt: –70 °C Density: 0.8279 g/cm³ (20 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064299 CAS Number: 1565-80-6 EINECS: 216-366-1 UN: 1105 ADR: 3,III Merck Index: 12,06109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...