(S)-(+)-2-Amino-3-methyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol
M-Clarity™ quality level = MQ200
186708-1G
186708-25G
186708-5G
SIAL186708-1GEA
60.2
GBP
SIAL186708-1G
SIAL186708-25G
SIAL186708-5G
(S)-(+)-2-Amino-3-methyl-1-butanol, Sigma-Aldrich®
(S)-(+)-2-Amino-3-methyl-1-butanol
Formula:
C₅H₁₃NO MW: 103.16 g/mol Boiling Pt: 187 °C (1013 hPa) Melting Pt: 31…32 °C Density: 0.92 g/cm³ (20 °C) |
MDL Number:
MFCD00064296 CAS Number: 2026-48-4 EINECS: 217-975-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...