L(+)-Valine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Val-OH, Valine, (S)-(+)-Valine

M-Clarity™ quality level = MQ200

V0500-100G V0500-1G V0500-1KG V0500-25G V0500-500G
SIALV0500-100GEA 138 GBP
SIALV0500-100G SIALV0500-1G SIALV0500-1KG SIALV0500-25G SIALV0500-500G
L(+)-Valine, Sigma-Aldrich®
L(+)-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR