L(+)-Valine

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Val-OH, Valine, (S)-(+)-Valine

38064.01 38064.02 38064.03
SERA38064.01EA 0 GBP
SERA38064.01 SERA38064.02 SERA38064.03
L(+)-Valine
L(+)-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217
Melting Pt: 86
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR