L(+)-Isoleucine

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

26540.02 26540.03
SERA26540.02EA 0 GBP
SERA26540.02 SERA26540.03
L(+)-Isoleucine
L(+)-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR